Gene Transcripts

Gene Symbol
GATA5
Entrez Gene ID
140628
Gene Name
GATA binding protein 5
Gene Aliases
GATAS, bB379O24.1
Chromosome Location
Chr. 20: 62463497 - 62475970 on Build GRCh38
UniGene ID
Hs.352250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080473.4 NP_536721.1 7 2575
XM_006723699.3 XP_006723762.1 7 2595
GenBank mRNA BC047790.1 - 7 2604

View Full Product Details