Gene Transcripts

Gene Symbol
Thbs2
Entrez Gene ID
21826
Gene Name
thrombospondin 2
Gene Aliases
TSP2, Thbs-2
Chromosome Location
Chr. 17: 14665500 - 14694262 on Build GRCm38
UniGene ID
Mm.26688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011581.3 NP_035711.2 22 4084
GenBank mRNA AK076503.1 - 3 1801
AK132508.1 - 22 3914
AK165881.1 - 4 701
BC031843.1 - 11 1869
BC053702.1 - 22 3886 30062369
L06421.1 - 1 183
L07803.1 - 22 3912

View Full Product Details