Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011731.3 | NP_035861.2 | 11 | 1990 | |
GenBank mRNA | AF075261.1 | - | 11 | 1814 | |
AK076806.1 | - | 12 | 1721 | ||
AK143893.1 | - | 11 | 1963 | ||
AK144038.1 | - | 11 | 1991 | ||
AK163340.1 | - | 12 | 1879 | ||
AK164706.1 | - | 11 | 1835 | ||
AK164713.1 | - | 9 | 3867 | ||
BC013484.1 | - | 11 | 1901 | 4242168 |