Gene Transcripts
- Gene Symbol
- Mtx2
- Entrez Gene ID
- 53375
- Gene Name
- metaxin 2
- Gene Aliases
- 1500012G02Rik
- Chromosome Location
- Chr. 2: 74825812 - 74876748 on Build GRCm38
- UniGene ID
- Mm.292613
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016804.4 |
NP_058084.3
|
8, 9 |
743 |
|
GenBank mRNA |
AB041562.1 |
- |
8, 9 |
735 |
|
AF053550.1 |
- |
8, 9 |
716 |
|
AK005233.1 |
- |
8, 9 |
675 |
|
AK083055.1 |
- |
|
705 |
|
AK087933.1 |
- |
8, 9 |
731 |
|
AK168000.1 |
- |
8, 9 |
736 |
|
AK168021.1 |
- |
8, 9 |
706 |
|
AK168045.1 |
- |
8, 9 |
735 |
|
AK169048.1 |
- |
8, 9 |
739 |
|
AK207939.1 |
- |
1, 2 |
53 |
|
AK212612.1 |
- |
1, 2 |
53 |
|
AK216047.1 |
- |
1, 2 |
53 |
|
BC006641.1 |
- |
8, 9 |
693 |
3483318 |
View Full Product Details