Gene Transcripts

Gene Symbol
Sit1
Entrez Gene ID
54390
Gene Name
suppression inducing transmembrane adaptor 1
Gene Aliases
Sit
Chromosome Location
Chr. 4: 43482083 - 43483709 on Build GRCm38
UniGene ID
Mm.31775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019436.3 NP_062309.2 5 799
GenBank mRNA AJ236881.1 - 5 775
AK079882.1 - 5 801
BC064055.1 - 5 812 576098

View Full Product Details