Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
56380
Gene Name
AT rich interactive domain 3B (BRIGHT-like)
Gene Aliases
Bdp, Dri2
Chromosome Location
Chr. 9: 57790353 - 57836847 on Build GRCm38
UniGene ID
Mm.425588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019689.2 NP_062663.1 1 368
GenBank mRNA AF116847.1 - 1 368
AK038068.1 - 2 449
AK148073.1 - 2 465
BC049776.1 - 2 590
BC145330.1 - 1 362
BC150755.1 - 1 362 9087666

View Full Product Details