Gene Transcripts

Gene Symbol
Tsga8
Entrez Gene ID
100502723
Gene Name
testis specific gene A8
Gene Aliases
Halap-X, Halapx
Chromosome Location
Chr. X: 83486674 - 83487924 on Build GRCm38
UniGene ID
Mm.383210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021898.2 NP_068698.1 2 407
GenBank mRNA AB032764.1 - 2 410
BC139062.1 - 2 241

View Full Product Details