Gene Transcripts
- Gene Symbol
- Lrp10
- Entrez Gene ID
- 65107
- Gene Name
- low-density lipoprotein receptor-related protein 10
- Gene Aliases
- Lrp9
- Chromosome Location
- Chr. 14: 54464147 - 54470292 on Build GRCm38
- UniGene ID
- Mm.472719
- Species
- Mus musculus
- Gene Symbol
- Stau1
- Entrez Gene ID
- 20853
- Gene Name
- staufen (RNA binding protein) homolog 1 (Drosophila)
- Gene Aliases
- 5830401L18Rik, AW549911, C85792, Stau
- Chromosome Location
- Chr. 2: 166947549 - 166996299 on Build GRCm38
- UniGene ID
- Mm.73276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK193147.1 |
- |
|
54 |
|
View Full Product Details