Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039169.1 | NP_001034258.1 | 2 | 635 | |
NM_001039170.1 | NP_001034259.1 | 2 | 184 | ||
NM_023314.3 | NP_075803.2 | 2 | 184 | ||
GenBank mRNA | AF068116.1 | - | 2 | 98 | |
AK017221.1 | - | 2 | 636 | ||
AK018516.1 | - | 2 | 182 | ||
AK032965.1 | - | 2 | 188 | ||
AK138055.1 | - | 2 | 123 | ||
AK150495.1 | - | 2 | 123 | ||
AK199854.1 | - | 2 | 166 | ||
BC028903.1 | - | 2 | 234 | 4166275 | |
BC045153.1 | - | 2 | 118 | 6441327 | |
BC049077.1 | - | 2 | 152 | 5708556 |