Gene Transcripts
- Gene Symbol
- Dock7
- Entrez Gene ID
- 67299
- Gene Name
- dedicator of cytokinesis 7
- Gene Aliases
- 3110056M06Rik, Gm430, m
- Chromosome Location
- Chr. 4: 98936659 - 99122605 on Build GRCm38
- UniGene ID
- Mm.260623
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290636.1 |
NP_001277565.1
|
47, 48 |
6323 |
|
NM_026082.5 |
NP_080358.3
|
46, 47 |
6233 |
|
GenBank mRNA |
AK013336.1 |
- |
5, 6 |
612 |
|
AK014226.1 |
- |
3, 4 |
436 |
|
AK122554.2 |
- |
16, 17 |
1967 |
|
BC024823.1 |
- |
17, 18 |
2335 |
|
BC024917.1 |
- |
11, 12 |
1488 |
|
BC055032.1 |
- |
6, 7 |
729 |
|
BC086672.1 |
- |
27, 28 |
3741 |
30647593 |
BC094420.1 |
- |
35, 36 |
4524 |
|
View Full Product Details