Gene Transcripts

Gene Symbol
Isl2
Entrez Gene ID
104360
Gene Name
insulin related protein 2 (islet 2)
Gene Aliases
3110001N10Rik
Chromosome Location
Chr. 9: 55538677 - 55547667 on Build GRCm38
UniGene ID
Mm.273996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027397.3 NP_081673.2 6 1649
GenBank mRNA AK013964.1 - 6 1592
BC120880.2 - 5 1329
BC148307.2 - 5 1334

View Full Product Details