Gene Transcripts

Gene Symbol
Arhgef25
Entrez Gene ID
52666
Gene Name
Rho guanine nucleotide exchange factor (GEF) 25
Gene Aliases
2410008H17Rik, D10Ertd610e, GEFT
Chromosome Location
Chr. 10: 127182521 - 127190088 on Build GRCm38
UniGene ID
Mm.272230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166413.1 NP_001159885.1 3 388
NM_028027.3 NP_082303.2 3 450
GenBank mRNA AF487515.1 - 3 416
AK010452.1 - 3 416
AK158684.1 - 3 388
AK194714.1 - 1 160
BC041113.1 - 3 432
CF724487.1 - 3 335

View Full Product Details