Gene Transcripts

Gene Symbol
Pop7
Entrez Gene ID
74097
Gene Name
processing of precursor 7, ribonuclease P family, (S. cerevisiae)
Gene Aliases
0610037N12Rik, AI852017, Rpp20
Chromosome Location
Chr. 5: 137501439 - 137502429 on Build GRCm38
UniGene ID
Mm.290242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028753.2 NP_083029.1 2 267
GenBank mRNA AK002782.1 - 2 269
AK158620.1 - 1 179
AK162268.1 - 2 272
BC010780.1 - 2 181 4216163

View Full Product Details