Gene Transcripts

Gene Symbol
Kctd18
Entrez Gene ID
51960
Gene Name
potassium channel tetramerisation domain containing 18
Gene Aliases
4932411A20Rik, 6530404F10Rik
Chromosome Location
Chr. 1: 57955101 - 57970084 on Build GRCm38
UniGene ID
Mm.339711
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159864.1 NP_001153336.1 4 674
GenBank mRNA AK032658.1 - 4 643
AK132699.1 - 4 643
BC075692.1 - 4 706 30631161

View Full Product Details