Gene Transcripts
- Gene Symbol
- Trim8
- Entrez Gene ID
- 93679
- Gene Name
- tripartite motif-containing 8
- Gene Aliases
- AA408830, BB006755, GERP, Rnf27
- Chromosome Location
- Chr. 19: 46501648 - 46516805 on Build GRCm38
- UniGene ID
- Mm.392177
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053100.2 |
NP_444330.2
|
1, 2 |
1361 |
|
GenBank mRNA |
AF281047.1 |
- |
1, 2 |
710 |
|
AK052763.1 |
- |
1, 2 |
96 |
|
AK155303.1 |
- |
1, 2 |
1308 |
|
BC029283.1 |
- |
1, 2 |
1295 |
|
BC037065.1 |
- |
1, 2 |
646 |
|
BC060094.1 |
- |
1, 2 |
591 |
|
BC082298.1 |
- |
1, 2 |
1236 |
|
BC152343.1 |
- |
1, 2 |
697 |
40093266 |
BC152344.1 |
- |
1, 2 |
697 |
40093269 |
View Full Product Details