Gene Transcripts
- Gene Symbol
- Fbxl20
- Entrez Gene ID
- 72194
- Gene Name
- F-box and leucine-rich repeat protein 20
- Gene Aliases
- 2610511F20Rik, 4632423N09Rik, AI849362, AL117906, C86145, Fbl2, Scr, mKIAA4147
- Chromosome Location
- Chr. 11: 98082554 - 98150440 on Build GRCm38
- UniGene ID
- Mm.218350
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC004087.1 |
- |
1 |
1607 |
|
View Full Product Details