Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138672.2 | NP_619613.2 | 3 | 439 | |
GenBank mRNA | AB052959.1 | - | 3 | 360 | |
AB052960.1 | - | 3 | 380 | ||
AB105392.1 | - | 3 | 376 | ||
AF290914.2 | - | 3 | 370 | ||
AK149309.1 | - | 3 | 394 | ||
AK159416.1 | - | 3 | 394 | ||
BC058607.1 | - | 3 | 372 |