Gene Transcripts

Gene Symbol
C7
Entrez Gene ID
730
Gene Name
complement component 7
Gene Aliases
Chromosome Location
Chr. 5: 40909497 - 40982939 on Build GRCh38
UniGene ID
Hs.78065
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000587.2 NP_000578.2 18 2989
GenBank mRNA BC063851.1 - 18 2970
J03507.1 - 17 2875

View Full Product Details