Gene Transcripts
- Gene Symbol
- Zfp33b
- Entrez Gene ID
- 69623
- Gene Name
- zinc finger protein 33B
- Gene Aliases
- 2310012P17Rik
- Chromosome Location
- Chr. 5: 104570873 - 104584620 on Build GRCm38
- UniGene ID
- Mm.69919
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC046593.1 |
- |
1 |
154 |
|
- Gene Symbol
- Zfp958
- Entrez Gene ID
- 233987
- Gene Name
- zinc finger protein 958
- Gene Aliases
- Chromosome Location
- Chr. 8: 4613170 - 4630231 on Build GRCm38
- UniGene ID
- Mm.102799
- Species
- Mus musculus
View Full Product Details