Gene Transcripts

Gene Symbol
Lrrk1
Entrez Gene ID
233328
Gene Name
leucine-rich repeat kinase 1
Gene Aliases
AW319595, C230002E15Rik, D130026O16Rik, mKIAA1790
Chromosome Location
Chr. 7: 66258745 - 66388341 on Build GRCm38
UniGene ID
Mm.33284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146191.3 NP_666303.3 32 5640
GenBank mRNA AK040153.1 - 12 2322
AK076432.1 - 5 967
AK081849.1 - 2 633
AK129441.1 - 29 5053
AK147469.1 - 32 5626
AK154395.1 - 6 1159
BC027199.1 - 2 508
BC066159.1 - 1 162
BC080819.1 - 1 267

View Full Product Details