Gene Transcripts

Gene Symbol
Prss50
Entrez Gene ID
235631
Gene Name
protease, serine 50
Gene Aliases
79H19C, Tsp50
Chromosome Location
Chr. 9: 110857899 - 110864629 on Build GRCm38
UniGene ID
Mm.102265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146227.4 NP_666339.2 3 899
GenBank mRNA AF481144.1 - 3 885
AK045168.1 - 3 900
BC150686.1 - 3 898 9087597

View Full Product Details