Gene Transcripts

Gene Symbol
Usp9y
Entrez Gene ID
107868
Gene Name
ubiquitin specific peptidase 9, Y chromosome
Gene Aliases
Dffry, Fafl2
Chromosome Location
Chr. Y: 1298961 - 1459782 on Build GRCm38
UniGene ID
Mm.347376
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148943.2 NP_683745.2 6 699
GenBank mRNA AJ307017.1 - 6 699

View Full Product Details