Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290565.1 | NP_001277494.1 | 3 | 457 | |
NM_153577.2 | NP_705805.1 | 3 | 457 | ||
GenBank mRNA | BC004761.1 | - | 3 | 406 | |
BC023803.1 | - | 3 | 444 | 5352953 | |
BC056649.1 | - | 3 | 396 | 5036575 | |
BE626968.1 | - | 3 | 382 |