Gene Transcripts
- Gene Symbol
- Poglut1
- Entrez Gene ID
- 224143
- Gene Name
- protein O-glucosyltransferase 1
- Gene Aliases
- 9630046K23Rik, Clp46, Ktelc1, Rumi
- Chromosome Location
- Chr. 16: 38525058 - 38550256 on Build GRCm38
- UniGene ID
- Mm.284366
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172380.4 |
NP_759012.1
|
1, 2 |
175 |
|
GenBank mRNA |
AK031608.1 |
- |
1, 2 |
114 |
|
AK035948.1 |
- |
1, 2 |
123 |
|
AK036224.1 |
- |
1, 2 |
105 |
|
AK041321.1 |
- |
1, 2 |
113 |
|
AK079886.1 |
- |
1, 2 |
144 |
|
AK215456.1 |
- |
1, 2 |
17 |
|
AK216766.1 |
- |
1, 2 |
17 |
|
BC026809.1 |
- |
1, 2 |
126 |
4205469 |
CB249259.1 |
- |
1, 2 |
176 |
|
View Full Product Details