Gene Transcripts
- Gene Symbol
- AP1G1
- Entrez Gene ID
- 164
- Gene Name
- adaptor related protein complex 1 gamma 1 subunit
- Gene Aliases
- ADTG, CLAPG1
- Chromosome Location
- Chr. 16: 71729000 - 71809073 on Build GRCh38
- UniGene ID
- Hs.461253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001030007.1 |
NP_001025178.1
|
4, 5 |
790 |
|
NM_001128.5 |
NP_001119.3
|
4, 5 |
790 |
|
GenBank mRNA |
AB015317.1 |
- |
4, 5 |
495 |
|
AK128078.1 |
- |
7, 8 |
1345 |
|
AK225906.1 |
- |
4, 5 |
549 |
|
AK293233.1 |
- |
4, 5 |
440 |
|
AK294546.1 |
- |
5, 6 |
643 |
|
AK299630.1 |
- |
5, 6 |
724 |
|
AK314484.1 |
- |
4, 5 |
795 |
|
BC036283.1 |
- |
4, 5 |
755 |
4158263 |
BU902252.1 |
- |
4, 5 |
790 |
|
Y12226.1 |
- |
4, 5 |
504 |
|
View Full Product Details