Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175448.3 | NP_780657.1 | 2 | 686 | |
GenBank mRNA | AK039308.1 | - | 2 | 687 | |
AK049896.1 | - | 2 | 247 | ||
AK049988.1 | - | 2 | 247 | ||
AK053797.1 | - | 2 | 709 | ||
AK139409.1 | - | 2 | 761 | ||
AK139868.1 | - | 2 | 739 | ||
AK163214.1 | - | 2 | 707 | ||
AK169009.1 | - | 2 | 856 | ||
BC058539.1 | - | 2 | 693 | 5690124 |