Gene Transcripts
- Gene Symbol
- Ttc16
- Entrez Gene ID
- 338348
- Gene Name
- tetratricopeptide repeat domain 16
- Gene Aliases
- 1200002K10Rik
- Chromosome Location
- Chr. 2: 32757026 - 32775663 on Build GRCm38
- UniGene ID
- Mm.475130
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290563.1 |
NP_001277492.1
|
13, 14 |
2018 |
|
NM_001290564.1 |
NP_001277493.1
|
13, 14 |
2073 |
|
NM_001290566.1 |
NP_001277495.1
|
12, 13 |
1771 |
|
NM_001290567.1 |
NP_001277496.1
|
13, 14 |
1819 |
|
NM_177384.3 |
NP_796358.2
|
13, 14 |
2018 |
|
GenBank mRNA |
AK028040.1 |
- |
13, 14 |
2073 |
|
AK089384.1 |
- |
10, 11 |
2874 |
|
AK133273.1 |
- |
13, 14 |
1796 |
|
AK160435.1 |
- |
13, 14 |
1769 |
|
BC140991.1 |
- |
12, 13 |
1762 |
9055603 |
BC145064.1 |
- |
13, 14 |
1810 |
9053612 |
View Full Product Details