Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPL
Entrez Gene ID
838144
Gene Name
Transducin family protein / WD-40 repeat family protein
Gene Aliases
AT1G15750, F7H2.9, F7H2_9, TOPLESS, WSIP1, WUS-INTERACTING PROTEIN 1
Chromosome Location
Chr.1: 5414817 - 5421033 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035973.3 NP_001031050.1 20 - 21 3513 91
NM_001035974.4 NP_001031051.1 21 - 22 3247 91
NM_101443.4 NP_563981.1 21 - 22 3407 91
NM_179341.3 NP_849672.1 22 - 23 3317 91
GenBank mRNA AY046013.1 - 21 - 22 3177 91
AY142619.1 - 20 - 21 3013 91

View Full Product Details