Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHCB6
Entrez Gene ID
838151
Gene Name
light harvesting complex photosystem II subunit 6
Gene Aliases
AT1G15820, CHLOROPHYLL PROTEIN 24, CP24, F7H2.16, F7H2_16, light harvesting complex photosystem II subunit 6
Chromosome Location
Chr.1: 5446462 - 5447880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101450.5 NP_173034.1 1 - 2 432 80
GenBank mRNA AF332425.1 - 1 - 2 228 80
AY065113.1 - 1 - 2 320 80
AY081644.1 - 1 - 2 228 80

View Full Product Details