Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATKDSA2
Entrez Gene ID
838203
Gene Name
Aldolase superfamily protein
Gene Aliases
AT1G16340, ATKSDA, AtKDO8PS, F3O9.14, F3O9_14
Chromosome Location
Chr.1: 5587722 - 5590761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035978.2 NP_001031055.1 5 - 6 405 87
NM_001084078.1 NP_001077547.1 5 - 6 345 87
NM_001198082.1 NP_001185011.1 4 - 5 481 87
NM_101500.4 NP_173084.1 4 - 5 594 87
GenBank mRNA AK175407.1 - 5 - 6 377 87
BT010438.1 - 4 - 5 302 87

View Full Product Details