Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATB BETA
Entrez Gene ID
838348
Gene Name
Protein phosphatase 2A, regulatory subunit PR55
Gene Aliases
AT1G17720, F11A6.6, F11A6_6
Chromosome Location
Chr.1: 6093185 - 6098443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101634.4 NP_564033.1 2 - 3 584 138
NM_179350.4 NP_849681.1 2 - 3 588 138
GenBank mRNA AF370519.1 - 2 - 3 478 138
AK226540.1 - 2 - 3 528 138
AY045665.1 - 2 - 3 475 138
BT000645.1 - 2 - 3 210 138
BT002183.1 - 2 - 3 210 138
U40161.1 - 2 - 3 296 138

View Full Product Details