Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDKD1;3
Entrez Gene ID
838384
Gene Name
cyclin-dependent kinase D1;3
Gene Aliases
AT1G18040, AT;CDCKD;3, CAK2AT, CYCLIN-DEPENDENT KINASE D1;3, T10F20.5, T10F20_5, cyclin-dependent kinase D1;3
Chromosome Location
Chr.1: 6206871 - 6209528 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101666.3 NP_173244.1 5 - 6 1105 119
GenBank mRNA AB047274.1 - 5 - 6 988 119
AY099677.1 - 5 - 6 1077 119
AY128857.1 - 5 - 6 876 119

View Full Product Details