Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF19
Entrez Gene ID
838505
Gene Name
auxin response factor 19
Gene Aliases
ARF11, AT1G19220, AUXIN RESPONSE FACTOR11, IAA22, T29M8.9, T29M8_9, auxin response factor 19, indole-3-acetic acid inducible 22
Chromosome Location
Chr.1: 6627683 - 6633133 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101780.4 NP_173356.1 5 - 6 816 57
GenBank mRNA AB493467.1 - 5 - 6 462 57
AY669794.1 - 5 - 6 462 57
U53672.2 - 5 - 6 680 57

View Full Product Details