The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335074.1 | NP_001325256.1 | 6 - 7 | 925 | 57 |
NM_126234.3 | NP_178282.2 | 7 - 8 | 839 | 57 | |
GenBank mRNA | AK221561.1 | - | 7 - 8 | 771 | 57 |