Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPSF73-II
Entrez Gene ID
814702
Gene Name
cleavage and polyadenylation specificity factor 73 kDa subunit-II
Gene Aliases
AT2G01730, ATCPSF73-II, CLEAVAGE AND POLYADENYLATION SPECIFICITY FACTOR 73 KDA SUBUNIT-II, EDA26, T8O11.10, T8O11_10, cleavage and polyadenylation specificity factor 73 kDa subunit-II, embryo sac development arrest 26
Chromosome Location
Chr.2: 320433 - 323963 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335074.1 NP_001325256.1 8 - 9 1056 115
NM_126234.3 NP_178282.2 9 - 10 970 115
GenBank mRNA AK221561.1 - 9 - 10 902 115

View Full Product Details