Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDD5
Entrez Gene ID
814738
Gene Name
indeterminate(ID)-domain 5
Gene Aliases
AT2G02070, AtIDD5, F5O4.16, F5O4_16, indeterminate(ID)-domain 5
Chromosome Location
Chr.2: 505375 - 510781 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335091.1 NP_001324413.1 2 - 3 724 75
NM_126268.3 NP_178316.1 2 - 3 724 75
GenBank mRNA AJ621495.1 - 2 - 3 576 75
AY056174.1 - 2 - 3 642 75
BT001215.1 - 2 - 3 576 75

View Full Product Details