Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD29
Entrez Gene ID
814808
Gene Name
IQ-domain 29
Gene Aliases
AT2G02790, IQ-domain 29, T20F6.7, T20F6_7
Chromosome Location
Chr.2: 787885 - 791289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335144.1 NP_001324495.1 3 - 4 990 81
NM_001335145.1 NP_001324494.1 3 - 4 533 81
NM_126334.3 NP_178382.2 3 - 4 517 81

View Full Product Details