Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTC4
Entrez Gene ID
821206
Gene Name
Inositol monophosphatase family protein
Gene Aliases
AT3G02870, F13E7.19, F13E7_19
Chromosome Location
Chr.3: 627512 - 629925 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035549.1 NP_001030626.1 4 - 5 358 113
NM_001125086.1 NP_001118558.1 4 - 5 349 113
NM_111155.3 NP_186936.1 4 - 5 438 113
GenBank mRNA AY035150.1 - 4 - 5 283 113
AY063021.1 - 4 - 5 195 113

View Full Product Details