Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPS4
Entrez Gene ID
814881
Gene Name
ureide permease 4
Gene Aliases
AT2G03520, ATUPS4, T4M8.4, T4M8_4, ureide permease 4
Chromosome Location
Chr.2: 1069491 - 1071421 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126402.3 NP_178450.2 6 - 7 1055 102

View Full Product Details