Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCG41
Entrez Gene ID
827186
Gene Name
pleiotropic drug resistance 13
Gene Aliases
AT4G15215, ATP-binding cassette G41, ATPDR13, PDR13, PLEIOTROPIC DRUG RESISTANCE 13, pleiotropic drug resistance 13
Chromosome Location
Chr.4: 8671349 - 8679176 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341000.1 NP_001329358.1 16 - 17 2393 98
NM_001341001.1 NP_001329357.1 15 - 16 2442 98
NM_001341002.1 NP_001329360.1 18 - 19 2772 98
NM_001341003.1 NP_001329359.1 16 - 17 2956 98
NM_001341004.1 NP_001329364.1 17 - 18 2880 98
NM_001341005.1 NP_001329363.1 17 - 18 2813 98
NM_001341006.1 NP_001329362.1 18 - 19 2720 98
NM_001341007.1 NP_001329361.1 17 - 18 2830 98
NM_001341008.1 NP_001329366.1 17 - 18 2763 98
NM_001341009.1 NP_001329365.1 17 - 18 2594 98
NM_148326.2 NP_680692.1 15 - 16 2355 98

View Full Product Details