Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE15
Entrez Gene ID
816200
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT2G16970, F12A24.15, F12A24_15, maternal effect embryo arrest 15
Chromosome Location
Chr.2: 7369452 - 7375688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335509.1 NP_001323497.1 10 - 11 1190 72
NM_001335510.1 NP_001323498.1 10 - 11 1190 72
NM_001335511.1 NP_001323499.1 10 - 11 1190 72
NM_001335512.1 NP_001323502.1 8 - 9 869 72
NM_001335513.1 NP_001323500.1 10 - 11 1190 72
NM_001335514.1 NP_001323501.1 10 - 11 1147 72

View Full Product Details