Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXL5
Entrez Gene ID
816228
Gene Name
EXORDIUM like 5
Gene Aliases
AT2G17230, EXORDIUM like 5, T23A1.9, T23A1_9
Chromosome Location
Chr.2: 7494536 - 7496292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127277.3 NP_565409.1 1 - 1 1233 75
GenBank mRNA AY058063.1 - 1 - 1 957 75
AY090303.1 - 1 - 1 924 75

View Full Product Details