Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MLO7
- Entrez Gene ID
-
816249
- Gene Name
- Seven transmembrane MLO family protein
- Gene Aliases
-
AT2G17430, ATMLO7, F5J6.19, F5J6_19, MILDEW RESISTANCE LOCUS O 7, NORTIA, NTA
- Chromosome Location
-
Chr.2: 7568018 - 7570912 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127298.5 |
NP_179335.3 |
8 - 9 |
929 |
81 |
View Full Product Details