Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRS4
Entrez Gene ID
816325
Gene Name
SHI-related sequence 4
Gene Aliases
AT2G18120, F8D23.10, F8D23_10, SHI-related sequence 4
Chromosome Location
Chr.2: 7876544 - 7878072 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127369.3 NP_179404.1 1 - 2 836 98

View Full Product Details