The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001338685.1 | NP_001326470.1 | 6 - 7 | 1394 | 88 |
NM_180299.1 | NP_850630.1 | 6 - 7 | 1252 | 88 |