Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPL2
Entrez Gene ID
831743
Gene Name
carboxyl-terminal domain (ctd) phosphatase-like 2
Gene Aliases
AT5G01270, ATCPL2, F7J8.250, F7J8_250, carboxyl-terminal domain (ctd) phosphatase-like 2
Chromosome Location
Chr.5: 107547 - 112324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203270.1 NP_001190199.1 11 - 12 2176 99
NM_120205.5 NP_195747.2 11 - 12 2189 99
GenBank mRNA AY557187.1 - 11 - 12 2008 99

View Full Product Details