Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PR-1-LIKE
Entrez Gene ID
816518
Gene Name
pathogenesis-related protein-1-like protein
Gene Aliases
AT2G19990, T2G17.21, T2G17_21, pathogenesis-related protein-1-like
Chromosome Location
Chr.2: 8629857 - 8630688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127557.3 NP_179589.1 1 - 1 537 86
GenBank mRNA BT011765.2 - 1 - 1 537 86
BT020497.1 - 1 - 1 444 86

View Full Product Details