Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NFXL2
- Entrez Gene ID
-
830450
- Gene Name
- NF-X1-type zinc finger protein NFXL2
- Gene Aliases
-
AT5G05660, ATNFXL2, Arabidopsis NF-X like 2, EARLY BIRD, EBI, MJJ3.6, MJJ3_6, NFX1-like 2
- Chromosome Location
-
Chr.5: 1690571 - 1695557 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120648.6 |
NP_196185.4 |
4 - 5 |
1492 |
77 |
View Full Product Details