Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPA70B
- Entrez Gene ID
-
830696
- Gene Name
- RPA70-kDa subunit B
- Gene Aliases
-
ARABIDOPSIS THALIANA RPA70-KDA SUBUNIT B, AT5G08020, ATRPA70B, F13G24.220, F13G24_220, RPA1B, RPA70-kDa subunit B, Replication Protein A 1B
- Chromosome Location
-
Chr.5: 2571957 - 2575061 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120884.2 |
NP_196419.1 |
1 - 2 |
278 |
87 |
View Full Product Details