The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335859.1 | NP_001324736.1 | 7 - 8 | 1534 | 93 |
NM_127927.3 | NP_565554.1 | 7 - 8 | 1430 | 93 |